Thermo Scientific Chemicals

Cyclopropanecarboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: L14931.06
5 g, Each
Thermo Scientific Chemicals

Cyclopropanecarboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: L14931.06
5 g, Each
Quantity
Catalog number: L14931.06
also known as L14931-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1489-69-6
IUPAC Name
cyclopropanecarbaldehyde
Molecular Formula
C4H6O
InChI Key
JMYVMOUINOAAPA-UHFFFAOYSA-N
SMILES
O=CC1CC1
Assay (GC)
≥97.5%
Refractive Index
1.4275-1.4315 @ 20?C
Appearance (Color)
Clear colorless
Free acid (titration)
≤1.5%
Form
Liquid

Description

Cyclopropanecarboxaldehyde reacts with Grignard or organolithium reagents to give the expected secondary alcohols. These are susceptible to ring opening in the presence of HBr, providing stereoselective access to homoallylic bromides. It is also used in the synthesis of eta(2)-enonenickel complexes by reacting with [Ni(cod)(2)] (cod = 1,5-cyclooctadiene) and PBu(3) and to prepare a cyclopropylidene tetralone which underwent an enantioselective palladium-catalyzed [3+2] TMM cycloaddition providing a chiral spiro-fused cyclopentene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropanecarboxaldehyde reacts with Grignard or organolithium reagents to give the expected secondary alcohols. These are susceptible to ring opening in the presence of HBr, providing stereoselective access to homoallylic bromides. It is also used in the synthesis of eta(2)-enonenickel complexes by reacting with [Ni(cod)(2)] (cod = 1,5-cyclooctadiene) and PBu(3) and to prepare a cyclopropylidene tetralone which underwent an enantioselective palladium-catalyzed [3+2] TMM cycloaddition providing a chiral spiro-fused cyclopentene.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. It is sensitive to air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text