Thermo Scientific Chemicals

Ethyl 4-hydroxy-3-methoxycinnamate, 98%, Thermo Scientific Chemicals

Catalog number: L14960.MD
250 mg, Each
Thermo Scientific Chemicals

Ethyl 4-hydroxy-3-methoxycinnamate, 98%, Thermo Scientific Chemicals

Catalog number: L14960.MD
250 mg, Each
Quantity
Catalog number: L14960.MD
also known as L14960-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4046-02-0
IUPAC Name
ethyl (2E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoate
Molecular Formula
C12H14O4
InChI Key
ATJVZXXHKSYELS-FNORWQNLSA-N
SMILES
CCOC(=O)\C=C\C1=CC=C(O)C(OC)=C1
Appearance (Color)
White to cream to pale orange to pale red
Assay (GC)
≥97.5%
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text