Thermo Scientific Chemicals

3-Methylxanthine, 98+%, Thermo Scientific Chemicals

Catalog number: L14986.MD
250 mg, Each
Thermo Scientific Chemicals

3-Methylxanthine, 98+%, Thermo Scientific Chemicals

Catalog number: L14986.MD
250 mg, Each
Quantity
Catalog number: L14986.MD
also known as L14986-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1076-22-8
IUPAC Name
3-methyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular Formula
C6H6N4O2
InChI Key
GMSNIKWWOQHZGF-UHFFFAOYSA-N
SMILES
CN1C2=C(NC=N2)C(=O)NC1=O
Assay (HPLC)
≥98.0%
Form
Powder
Identification (FTIR)
Conforms
Appearance (Color)
Pale cream to cream

Description

3-Methylxanthine is a Xanthine derivative with diuretic, cardiac stimulant and smooth muscle relaxant activities; isomeric with theobromine.They can also act as Bronchodilator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylxanthine is a Xanthine derivative with diuretic, cardiac stimulant and smooth muscle relaxant activities; isomeric with theobromine.They can also act as Bronchodilator.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text