Thermo Scientific Chemicals

Hydroxyacetone, 95%, Thermo Scientific Chemicals

Catalog number: L15008.22
100 g, Each
Thermo Scientific Chemicals

Hydroxyacetone, 95%, Thermo Scientific Chemicals

Catalog number: L15008.22
100 g, Each
Quantity
Catalog number: L15008.22
also known as L15008-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
116-09-6
IUPAC Name
1-hydroxypropan-2-one
Molecular Formula
C3H6O2
InChI Key
XLSMFKSTNGKWQX-UHFFFAOYSA-N
SMILES
CC(=O)CO
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥94.0%
Refractive Index
1.4220-1.4280 @ 20?C

Description

Hydroxyacetone is used as a reagent in organic chemical reactions. It also serves as a component for Mannich reaction and aldol reactions. It is also used in the syntheses of 2-oxo-propionaldehyde, imidazoles, polyols, acrolein, dyes and skin tanning agents. It yields (R)-1,2-propanediol upon reduction of hydroxyacetone in the presence of a microbial cell catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydroxyacetone is used as a reagent in organic chemical reactions. It also serves as a component for Mannich reaction and aldol reactions. It is also used in the syntheses of 2-oxo-propionaldehyde, imidazoles, polyols, acrolein, dyes and skin tanning agents. It yields (R)-1,2-propanediol upon reduction of hydroxyacetone in the presence of a microbial cell catalyst.

Solubility
Miscible with water, alcohol and ether.

Notes
Store in cool place. Air sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text