Thermo Scientific Chemicals

Methacrylamide, 98%, Thermo Scientific Chemicals

Catalog number: L15013.36
500 g, Each
Thermo Scientific Chemicals

Methacrylamide, 98%, Thermo Scientific Chemicals

Catalog number: L15013.36
500 g, Each
Quantity
Catalog number: L15013.36
also known as L15013-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79-39-0
IUPAC Name
2-methylprop-2-enamide
Molecular Formula
C4H7NO
InChI Key
FQPSGWSUVKBHSU-UHFFFAOYSA-N
SMILES
CC(=C)C(N)=O
Appearance (Color)
White to pale yellow
Form
Crystalline powder or crystals
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
107-117?C

Description

Methacrylamide is a reagent used in the production of polymers and copolymers. It is also used as a raw material in the production of chemicals for industrial and professional use such as textiles, leather, fur, bulk, large scale chemicals (including petroleum products), fine chemicals, formulation [mixing] of preparations and/or re-packaging (excluding alloys), building and construction work, electricity, steam, gas, water supply and sewage treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methacrylamide is a reagent used in the production of polymers and copolymers. It is also used as a raw material in the production of chemicals for industrial and professional use such as textiles, leather, fur, bulk, large scale chemicals (including petroleum products), fine chemicals, formulation [mixing] of preparations and/or re-packaging (excluding alloys), building and construction work, electricity, steam, gas, water supply and sewage treatment.

Solubility
Soluble in water (202g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text