Thermo Scientific Chemicals

Polysorbate 20, Thermo Scientific Chemicals

Catalog number: L15029.AP
500 mL, Each
Thermo Scientific Chemicals

Polysorbate 20, Thermo Scientific Chemicals

Catalog number: L15029.AP
500 mL, Each
Quantity
Catalog number: L15029.AP
also known as L15029-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9005-64-5
Molecular Formula
(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
InChI Key
HMFKFHLTUCJZJO-UHFFFAOYNA-N
SMILES
CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
MDL Number
MFCD00165986
Appearance (Color)
Clear colorless or pale yellow to yellow
Form
Liquid
Hydroxyl Value
96-108 mg KOH/g
Refractive Index
1.4650-1.4750 @ 20°C

Description

Used for solubilizing membrane proteins during isolation and purification.Polysorbate 20 is used as a wetting agent in flavored mouth drops. It acts as a washing agent in Western blots and ELISAs and prevents antibody binding. It is a solubilizing agent of membrane proteins. In pharmaceutical chemistry, it is used as an excipient to stabilize emulsions and suspensions. Further, it is used as a dispersing agent, stabilizer, lubricator and also used in creams, salves, ointments, protective creams, balms, pomades, lipsticks, mascaras and glosses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text