Polysorbate 40, Thermo Scientific Chemicals
Polysorbate 40, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polysorbate 40, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number L15030.AP
also known as L15030-AP
Price (USD)/ Each
38.65
Online exclusive
43.00 
Save 4.35 (10%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
38.65
Online exclusive
43.00 
Save 4.35 (10%)
Add to cart
Polysorbate 40, Thermo Scientific Chemicals
Catalog numberL15030.AP
Price (USD)/ Each
38.65
Online exclusive
43.00 
Save 4.35 (10%)
-
Add to cart
Chemical Identifiers
CAS9005-66-7
Molecular Formula(C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6
InChI KeyDVRFSZFBSOKHQW-UHFFFAOYNA-N
SMILESCCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
MDL NumberMFCD00165345
SpecificationsSpecification SheetSpecification Sheet
Residue on ignition≤0.25%
Appearance (Color)Pale yellow to yellow
FormViscous oily liquid
Acid Value≤2.0%
CommentSaponification value 41-60
View more
Polysorbate 40 has been used in a study to assess the determination of iodide in urine based on a chemiluminescence system. It has also been used in a study to investigate the delivery of astaxanthin and canthaxanthin to HepG2 cells. Also used as oil-in-water emulsifiers in pharmaceuticals, cosmetics, cleaning compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only