Polysorbate 60, Thermo Scientific Chemicals
Polysorbate 60, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polysorbate 60, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number L15031.AE
also known as L15031-AE
Price (USD)/ Each
26.30
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
26.30
Add to cart
Polysorbate 60, Thermo Scientific Chemicals
Catalog numberL15031.AE
Price (USD)/ Each
26.30
-
Add to cart
Chemical Identifiers
CAS9005-67-8
Molecular Formula(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC24H46O6
InChI KeyDEKFZWMENCCOFU-UHFFFAOYNA-N
SMILESCCCCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
MDL NumberMFCD01780110
SpecificationsSpecification SheetSpecification Sheet
FormViscous liquid or gel or semi-solid or paste
Hydroxyl Value81.0-96.0 mgKOH/g
Acid Value≤ 2.2 mgKOH/g
Appearance (Color)Pale yellow to yellow to pale brown
Water Content (Karl Fischer Titration)≤ 3.0%
View more
Polysorbate 60 is used in food, medicine, plastic, cosmetics and other industries. It is an excellent emulgator and one of elements to constitute polyacrylonitrile fiber spinning oil. It also used as softening agent after processing fiber and eliminate fiber static and improve its softness.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only