Thermo Scientific Chemicals

Pyridine-3-boronic acid, Thermo Scientific Chemicals

Catalog number: L15040.03
1 g, Each
Thermo Scientific Chemicals

Pyridine-3-boronic acid, Thermo Scientific Chemicals

Catalog number: L15040.03
1 g, Each
Quantity
Catalog number: L15040.03
also known as L15040-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1692-25-7
IUPAC Name
(pyridin-3-yl)boronic acid
Molecular Formula
C5H6BNO2
InChI Key
ABMYEXAYWZJVOV-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=CN=C1
Appearance (Color)
White
Proton NMR
Conforms to structure
Form
Powder

Description

Reactant used for preparation of phosphine-free Suzuki-Miyaura cross-coupling reactions, regioselective Suzuki-Miyaura coupling and tandem palladium-catalyzed intermolecular aminocarbonylation and annulation, N-arylation using copper acetylacetonate catalys, copper-mediated cyanation and regioselective cyanation of electron-rich benzenes, New linear poly(phenylpyridyl) chains by Suzuki coupling and Oligopyridyl foldamers as mimics of a-helix twist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant used for preparation of phosphine-free Suzuki-Miyaura cross-coupling reactions, regioselective Suzuki-Miyaura coupling and tandem palladium-catalyzed intermolecular aminocarbonylation and annulation, N-arylation using copper acetylacetonate catalys, copper-mediated cyanation and regioselective cyanation of electron-rich benzenes, New linear poly(phenylpyridyl) chains by Suzuki coupling and Oligopyridyl foldamers as mimics of a-helix twist.

Solubility
Slightly soluble in methanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text