Thermo Scientific Chemicals

4',5,7-Trihydroxyflavone, 97%, Thermo Scientific Chemicals

Catalog number: L15041.MC
100 mg, Each
Thermo Scientific Chemicals

4',5,7-Trihydroxyflavone, 97%, Thermo Scientific Chemicals

Catalog number: L15041.MC
100 mg, Each
Quantity
Catalog number: L15041.MC
also known as L15041-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
520-36-5
IUPAC Name
5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one
Molecular Formula
C15H10O5
InChI Key
KZNIFHPLKGYRTM-UHFFFAOYSA-N
SMILES
OC1=CC=C(C=C1)C1=CC(=O)C2=C(O)C=C(O)C=C2O1
Appearance (Color)
Cream to yellow to brown
Assay (HPLC)
≥96.0%
Melting Point (clear melt)
295.0-305.0?C
Form
Powder

Description

4',5,7-Trihydroxyflavone is a nonmutagenic flavonoid shown to inhibit cell proliferation, angiogenesis and protein kinase. Also induces apoptosis in breast cancer cells. It inhibit topoisomerase I-catalyzed DNA re-ligation and enhance gap junctional intercellular communication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′,5,7-Trihydroxyflavone is a nonmutagenic flavonoid shown to inhibit cell proliferation, angiogenesis and protein kinase. Also induces apoptosis in breast cancer cells. It inhibit topoisomerase I-catalyzed DNA re-ligation and enhance gap junctional intercellular communication.

Solubility
Soluble in dimethyl sulfoxide (27 mg/ml) or 1M potassium hydroxide (50mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text