Thermo Scientific Chemicals

3-Carboxy-PROXYL, free radical, 97+%, Thermo Scientific Chemicals

Catalog number: L15068.MF
50 mg, Each
Thermo Scientific Chemicals

3-Carboxy-PROXYL, free radical, 97+%, Thermo Scientific Chemicals

Catalog number: L15068.MF
50 mg, Each
Quantity
Catalog number: L15068.MF
also known as L15068-MF
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2154-68-9
IUPAC Name
(3R)-1-hydroxy-2,2,5,5-tetramethylpyrrolidine-3-carboxylate
Molecular Formula
C9H16NO3
InChI Key
CLKPVQZFNYXFCY-LURJTMIESA-M
SMILES
CC1(C)C[C@@H](C([O-])=O)C(C)(C)N1O
Appearance (Color)
Pale yellow to yellow
Assay (HPLC)
≥97.0%
Form
Crystals or powder or crystalline powder

Description

3-Carboxy-PROXYL is used in the synthesis and reactions of stable nitroxyl radicals. It is also used to study molecular migration and long-term stability of seeds and pollen and used in the preparation of a tetraphenylporphyrin bearing a nitroxyl radical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Carboxy-PROXYL is used in the synthesis and reactions of stable nitroxyl radicals. It is also used to study molecular migration and long-term stability of seeds and pollen and used in the preparation of a tetraphenylporphyrin bearing a nitroxyl radical.

Solubility
Soluble in water (partly miscible), methanol (50 mg/ml), acetone, dichloromethane and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text