Thermo Scientific Chemicals

Acrylamide, electrophoresis grade, 99+%

Catalog number: L15075.36
500 g, Each
Thermo Scientific Chemicals

Acrylamide, electrophoresis grade, 99+%

Catalog number: L15075.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L15075.36
also known as L15075-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79-06-1
IUPAC Name
prop-2-enamide
Molecular Formula
C3H5NO
InChI Key
HRPVXLWXLXDGHG-UHFFFAOYSA-N
SMILES
NC(=O)C=C
Appearance (Color)
White
Form
Crystalline powder
Assay (GC)
> 99.0%

Description

Acrylamide is used as a monomer in the synthesis of polyacrylamides, which finds application as water-soluble thickeners in waste water treatment, gel electrophoresis, ore processing and dyes production. In molecular biology, it is used as a carrier as well as involved in the precipitation of small amounts of DNA. It plays an important role as an intermediate in the preparation of N-methylol acrylamide and N-butoxyacrylamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• Acrylamide / N,N'-Methylenebisacrylamide is a water-soluble, odorless, white crystalline powder
• This compound polymerizes in the presence of free radicals to generate gels whose pore-size may be regulated by manipulating the relative concentrations of this mixture of compounds
Applications
• Polyacrylamide gels are widely used in biochemistry, forensic chemistry, genetics, molecular biology, and biotechnology to separate proteins or nucleic acids, according to their electrophoretic mobility
• Because of their higher resolution compared to other gels, polyacrylamide gels are also used in the analysis of smaller RNA species from mixed samples
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text