Thermo Scientific Chemicals

1-Hexadecanethiol, 97% (dry wt.), may cont. up to 4% water, Thermo Scientific Chemicals

Catalog number: L15099.AE
100 mL, Each
Thermo Scientific Chemicals

1-Hexadecanethiol, 97% (dry wt.), may cont. up to 4% water, Thermo Scientific Chemicals

Catalog number: L15099.AE
100 mL, Each
Quantity
Catalog number: L15099.AE
also known as L15099-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2917-26-2
IUPAC Name
hexadecane-1-thiol
Molecular Formula
C16H34S
InChI Key
ORTRWBYBJVGVQC-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCS
Appearance (Color)
Clear to slightly hazy, colorless
Form
Liquid or low melting solid
Assay (GC)
≥96.0%
Comment
Material Sourced in UK and US
Water Content (Karl Fischer Titration)
≤4.0% (UK Sourced Material)

Description

1-Hexadecanethiol is used in the study of nanoparticles and self-assembled monolayers. It is also used to prepare thiol-functionalized 1.5 nm gold nanoparticles via a ligand exchange process with triphenylphosphine-stabilized nanoparticles and in the formation of hydrophobic self-assembled monolayers (SAMs). Due to the length of the alkane chain, the resulting monolayer is highly ordered. 1-Hexadecanethiol is also employed in different fields such as biomedicine, chemical industry, also used in electroplating industry and chemical auxiliaries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexadecanethiol is used in the study of nanoparticles and self-assembled monolayers. It is also used to prepare thiol-functionalized 1.5 nm gold nanoparticles via a ligand exchange process with triphenylphosphine-stabilized nanoparticles and in the formation of hydrophobic self-assembled monolayers (SAMs). Due to the length of the alkane chain, the resulting monolayer is highly ordered. 1-Hexadecanethiol is also employed in different fields such as biomedicine, chemical industry, also used in electroplating industry and chemical auxiliaries.

Solubility
Soluble in methanol, benzene, ether, acetone and ethyl acetate. Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store under inert gas. It is sensitive to air. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text