Thermo Scientific Chemicals

Diethylmethoxyborane, 1M soln in THF, Thermo Scientific Chemicals

Catalog number: L15107.AC
25 mL, Each
Thermo Scientific Chemicals

Diethylmethoxyborane, 1M soln in THF, Thermo Scientific Chemicals

Catalog number: L15107.AC
25 mL, Each
Quantity
Catalog number: L15107.AC
also known as L15107-AC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7397-46-8
IUPAC Name
methyl diethylborinate
Molecular Formula
C5H13BO
InChI Key
FESAXEDIWWXCNG-UHFFFAOYSA-N
SMILES
CCB(CC)OC
Appearance (Color)
Clear, colorless
Form
Liquid
Concentration
1.0 - 1.1 Molar

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text