Thermo Scientific Chemicals

Triethylborane, 1M soln. in THF, Thermo Scientific Chemicals

Catalog number: L15109.AE
100 mL, Each
Thermo Scientific Chemicals

Triethylborane, 1M soln. in THF, Thermo Scientific Chemicals

Catalog number: L15109.AE
100 mL, Each
Quantity
Catalog number: L15109.AE
also known as L15109-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
97-94-9
IUPAC Name
triethylborane
Molecular Formula
C6H15B
InChI Key
LALRXNPLTWZJIJ-UHFFFAOYSA-N
SMILES
CCB(CC)CC
Form
Liquid
Assay from Supplier's CofA
0.95-1.30 M
Appearance (Color)
Clear colorless to yellow to orange

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Triethylborane is catalyst for allylation of aldehydes, decarboxylative C-C bond cleavage reactions, rhenium hydride / boron Lewis acid cocatalysis of alkene hydrogenations, Regioselective hydroxyalkylation of unsaturated oxime ethers. Reactant for radical reductions of alkyl bromides with N-heterocyclic carbene boranes and synthesis of tetramethylammonium trialkylphenylborate salts with oxidation potential.

Solubility
Reacts with water.

Notes
Air Sensitive, store away form air. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text