Thermo Scientific Chemicals

Borane-morpholine complex, 97%, Thermo Scientific Chemicals

Catalog number: L15110.06
5 g, Each
Thermo Scientific Chemicals

Borane-morpholine complex, 97%, Thermo Scientific Chemicals

Catalog number: L15110.06
5 g, Each
Quantity
Catalog number: L15110.06
also known as L15110-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4856-95-5
IUPAC Name
morpholine borane
Molecular Formula
C4H12BNO
InChI Key
IFVAHVOGOBRFSP-UHFFFAOYSA-N
SMILES
B.C1COCCN1
Appearance (Color)
White
Assay (special)
≥95.0% (EA)
Elemental Analysis
Nitrogen, N: 13.18-14.57% (Theory: 13.87%)
Elemental Analysis
Hydrogen, H: 11.38-12.58% (Theory: 11.98%)
Elemental Analysis
Carbon, C: 45.21-49.97% (Theory: 47.59%)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text