Borane-morpholine complex, 97%, Thermo Scientific Chemicals
Borane-morpholine complex, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Borane-morpholine complex, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L15110.14
also known as L15110-14
Price (USD)/ Each
84.65
Online exclusive
94.10 
Save 9.45 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
84.65
Online exclusive
94.10 
Save 9.45 (10%)
Add to cart
Borane-morpholine complex, 97%, Thermo Scientific Chemicals
Catalog numberL15110.14
Price (USD)/ Each
84.65
Online exclusive
94.10 
Save 9.45 (10%)
-
Add to cart
Chemical Identifiers
CAS4856-95-5
IUPAC Namemorpholine borane
Molecular FormulaC4H12BNO
InChI KeyIFVAHVOGOBRFSP-UHFFFAOYSA-N
SMILESB.C1COCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Elemental AnalysisBoron, B: 10.17-11.25% (Theory: 10.71%)
Appearance (Color)White
Assay (special)≥95.0% (EA)
Elemental AnalysisNitrogen, N: 13.18-14.57% (Theory: 13.87%)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only