Thermo Scientific Chemicals

Sodium tert-pentyloxide, 95%, Thermo Scientific Chemicals

Catalog number: L15112.22
100 g, Each
Thermo Scientific Chemicals

Sodium tert-pentyloxide, 95%, Thermo Scientific Chemicals

Catalog number: L15112.22
100 g, Each
Quantity
Catalog number: L15112.22
also known as L15112-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14593-46-5
IUPAC Name
sodium 2,2-dimethylpropan-1-olate
Molecular Formula
C5H11NaO
InChI Key
YQMLGVNRTAQUFQ-UHFFFAOYSA-N
SMILES
[Na+].CC(C)(C)C[O-]
Appearance (Color)
White to yellow to brown
Form
Powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥94.0%

Description

Sodium tert-pentyloxide is a effective base for the alkylation of ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium tert-pentyloxide is a effective base for the alkylation of ketones.

Solubility
Reacts with water.

Notes
Air and Moisture Sensitive, store away form air and water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text