3-Iodophenylacetonitrile, 97%, Thermo Scientific Chemicals
3-Iodophenylacetonitrile, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Iodophenylacetonitrile, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L15118.03
also known as L15118-03
Price (USD)/ Each
69.30
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
69.30
Add to cart
3-Iodophenylacetonitrile, 97%, Thermo Scientific Chemicals
Catalog numberL15118.03
Price (USD)/ Each
69.30
-
Add to cart
Chemical Identifiers
CAS130723-54-5
IUPAC Name2-(3-iodophenyl)acetonitrile
Molecular FormulaC8H6IN
InChI KeyLVOKGAHCTHNWIL-UHFFFAOYSA-N
SMILESIC1=CC=CC(CC#N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to pale orange
FormCrystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)≥96.0%
Melting Point (clear melt)31.5-37.5?C
3-Iodophenylacetonitrile is a important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodophenylacetonitrile is a important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to light. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Xing Fu.; Ping-Zhong Tan.; Nora S. Kula.; Ross Baldessarini.; Gilles Tamagnan.; Robert B. Innis.; Ronald M. Baldwin. Synthesis, Receptor Potency, and Selectivity of Halogenated Diphenylpiperidines as Serotonin 5-HT2A Ligands for PET or SPECT Brain Imaging. J. Med. Chem. 2002, 45 (11), 2319-2324.
  2. Takashi Mano.; Rodney W. Stevens.; Yoshiyuki Okumura.; Makoto Kawai.; Takako Okumura.; Minoru Sakakibara. 5-Lipoxygenase inhibitors: convenient synthesis of 4-[3-(4-heterocyclylphenylthio)phenyl]-3,4,5,6-tetrahydro-2H-pyran-4-carboxamide analogues. Bioorganic & Medicinal Chemistry Letters. 2005, 15 (10), 2611-2615.