Thermo Scientific Chemicals

3-Iodophenylacetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: L15118.06
5 g, Each
Thermo Scientific Chemicals

3-Iodophenylacetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: L15118.06
5 g, Each
Quantity
Catalog number: L15118.06
also known as L15118-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
130723-54-5
IUPAC Name
2-(3-iodophenyl)acetonitrile
Molecular Formula
C8H6IN
InChI Key
LVOKGAHCTHNWIL-UHFFFAOYSA-N
SMILES
IC1=CC=CC(CC#N)=C1
Appearance (Color)
White to yellow to pale orange
Form
Crystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)
≥96.0%
Melting Point (clear melt)
31.5-37.5?C

Description

3-Iodophenylacetonitrile is a important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodophenylacetonitrile is a important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to light. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text