Thermo Scientific Chemicals

4-Iodophenylacetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: L15119.06
5 g, Each
Thermo Scientific Chemicals

4-Iodophenylacetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: L15119.06
5 g, Each
Quantity
Catalog number: L15119.06
also known as L15119-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51628-12-7
IUPAC Name
2-(4-iodophenyl)acetonitrile
Molecular Formula
C8H6IN
InChI Key
PNXWQTYSBFGIFD-UHFFFAOYSA-N
SMILES
IC1=CC=C(CC#N)C=C1
Assay (GC)
> 96.0%
Appearance (Color)
White to cream or yellow
Form
Crystalline or fused solid or powder

Description

4-Iodophenylacetonitrile is used as an intermediate in organic syntheses and in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodophenylacetonitrile is used as an intermediate in organic syntheses and in pharmaceuticals.

Solubility
Insoluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text