Thermo Scientific Chemicals

3-Thiopheneethanol, 98%, Thermo Scientific Chemicals

Catalog number: L15138.06
5 g, Each
Thermo Scientific Chemicals

3-Thiopheneethanol, 98%, Thermo Scientific Chemicals

Catalog number: L15138.06
5 g, Each
Quantity
Catalog number: L15138.06
also known as L15138-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13781-67-4
IUPAC Name
2-(thiophen-3-yl)ethan-1-ol
Molecular Formula
C6H8OS
InChI Key
YYPNNBPPDFTQFX-UHFFFAOYSA-N
SMILES
OCCC1=CSC=C1
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.5500-1.5545 @ 20°C
Appearance (Color)
Clear colorless to dark yellow
Assay (GC)
≥97.5%

Description

3-Thiopheneethanol is used in the synthesis of various ether and ester derivatives such as 3-(2-(benzyloxy)ethyl)thiophene, 3-[2-((triphenylmethyl)oxy)ethyl]thiophene, 3-[2-((trimethylsilyl)oxy)ethyl]thiophene, 3-[2-((dimethyl-tert-butylsilyl)oxy)ethyl]thiophene, 3-(2-acetoxyethyl)thiophene and 3-(2-(benzoyloxy)ethyl)thiophene

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Thiopheneethanol is used in the synthesis of various ether and ester derivatives such as 3-(2-(benzyloxy)ethyl)thiophene, 3-[2-((triphenylmethyl)oxy)ethyl]thiophene, 3-[2-((trimethylsilyl)oxy)ethyl]thiophene, 3-[2-((dimethyl-tert-butylsilyl)oxy)ethyl]thiophene, 3-(2-acetoxyethyl)thiophene and 3-(2-(benzoyloxy)ethyl)thiophene

Solubility
Sparingly soluble in water(0.059 g/L at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text