Thermo Scientific Chemicals

(tert-Butoxycarbonylmethylene)triphenylphosphorane, 98%, Thermo Scientific Chemicals

Catalog number: L15144.14
25 g, Each
Thermo Scientific Chemicals

(tert-Butoxycarbonylmethylene)triphenylphosphorane, 98%, Thermo Scientific Chemicals

Catalog number: L15144.14
25 g, Each
Quantity
Catalog number: L15144.14
also known as L15144-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
35000-38-5
IUPAC Name
tert-butyl 2-(triphenyl-λ⁵-phosphanylidene)acetate
Molecular Formula
C24H25O2P
InChI Key
ZWZUFQPXYVYAFO-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Assay (HPLC)
≥97.5%
Appearance (Color)
White to pale yellow
Form
Crystals or powder or crystalline powder

Description

(tert-Butoxycarbonylmethylene)triphenylphosphorane is a chemical used in the synthesis of aldose reductase inhibitors, a podophyllotoxin derivative and tautomycin, a podophyllotoxin derivative and tautomycin. It Is also used in medicinal chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(tert-Butoxycarbonylmethylene)triphenylphosphorane is a chemical used in the synthesis of aldose reductase inhibitors, a podophyllotoxin derivative and tautomycin, a podophyllotoxin derivative and tautomycin. It Is also used in medicinal chemistry.

Solubility
Sparingly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text