(-)-Fenchone, 98+%, Thermo Scientific Chemicals
(-)-Fenchone, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(-)-Fenchone, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number L15151.30
also known as L15151-30
Price (USD)/ Each
87.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
87.00
Add to cart
(-)-Fenchone, 98+%, Thermo Scientific Chemicals
Catalog numberL15151.30
Price (USD)/ Each
87.00
-
Add to cart
Chemical Identifiers
CAS7787-20-4
IUPAC Name(1R,4S)-1,3,3-trimethylbicyclo[2.2.1]heptan-2-one
Molecular FormulaC10H16O
InChI KeyLHXDLQBQYFFVNW-OIBJUYFYSA-N
SMILESCC1(C)[C@H]2CC[C@](C)(C2)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Optical Rotation-52.8 ± 2.0? (neat)
Refractive Index1.4605-1.4645 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only