Rhodium(II) acetate, dimer, 98+%, Thermo Scientific Chemicals
Rhodium(II) acetate, dimer, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Rhodium(II) acetate, dimer, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Catalog number L15152.MC
Price (USD)/ Each
-
Quantity:
100 mg
Request bulk or custom format
Chemical Identifiers
CAS15956-28-2
IUPAC Nametetramethyl-2,4,6,8,9,11,12,14-octaoxa-1,5-dirhodatetracyclo[3.3.3.3¹,⁵.0¹,⁵]tetradecane-1,1,5,5-tetrakis(ylium)-3,7,10,13-tetraide
Molecular FormulaC8H12O8Rh2
InChI KeyVOXKIADTKKPBBP-UHFFFAOYSA-N
SMILESC[C-]1O[Rh++]234O[C-](C)O[Rh++]2(O1)(O[C-](C)O3)O[C-](C)O4
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green
Assay (unspecified)(by Elemental Rhodium): >98.0 to <102.0%
FormCrystalline powder
Elemental Analysis(Rhodium, Rh): 45.63-47.49%
Catalyst for cyclopropanation of alkenes, oxidation of alcohols, cyclization reactions involving ?-diazo carbonyl groups, insertion into C-H and X-H bonds (X-H is NH or SH or OH) and for ylide formation. More reactive and useful in differentiating ribonucleosides and deoxynucleosides. It is also used in functionalizing fullerenes into polymers. Efficient catalyst for hydrogen transfer from 2-propanol to cyclohexanone and other unsaturated compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Catalyst for cyclopropanation of alkenes, oxidation of alcohols, cyclization reactions involving ɑ-diazo carbonyl groups, insertion into C-H and X-H bonds (X-H is NH or SH or OH) and for ylide formation. More reactive and useful in differentiating ribonucleosides and deoxynucleosides. It is also used in functionalizing fullerenes into polymers. Efficient catalyst for hydrogen transfer from 2-propanol to cyclohexanone and other unsaturated compounds.

Solubility
Soluble in aqueous solution like water. Slightly soluble in ethanol

Notes
Stable under normal conditions.
RUO – Research Use Only

General References:

  1. Catalyst for various cyclization reactions involving ɑ-diazo carbonyl groups. Promotes the formation of macrocyclic lactones via intramolecular cyclopropanation and carbon-hydrogen insertion: J. Am. Chem. Soc., 117, 7181 (1995). For a brief feature on uses of the reagent in synthesis, see: Synlett, 3169 (2005).
  2. Aghabali, A.; Olmstead, M. M. Zipping up Fullerenes into Polymers Using Rhodium(II) Acetate Dimer and N(CH2CH2)2NC60 as Building Blocks. Chem. Commun. (Camb). 2014, 50 (96), 15152-15155.
  3. Honey, M. A.; Moody, C. J. Synthesis of Indoxylic Acid Esters by Rhodium-catalyzed Carbene N-H Insertion and Thermal Cyclization. Australian J. Chem. 2014, 67 (9), 1211-1216.