Thermo Scientific Chemicals

Montmorillonite K10, Thermo Scientific Chemicals

Catalog number: L15160.30
250 g, Each
Thermo Scientific Chemicals

Montmorillonite K10, Thermo Scientific Chemicals

Catalog number: L15160.30
250 g, Each
Quantity
Catalog number: L15160.30
also known as L15160-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1318-93-0
Appearance (Color)
White to cream or pale brown or pale grey
Application test
Confirms K10 grade
Form
Powder or granules

Description

Used for support for catalysts, preparation of clay nanopaper composites of nacre-like structure, conversion of triose sugars with alcohols to alkyl lactates and Deprotection of ketone dimethylhydrazones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used for support for catalysts, preparation of clay nanopaper composites of nacre-like structure, conversion of triose sugars with alcohols to alkyl lactates and Deprotection of ketone dimethylhydrazones.

Solubility
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text