Thermo Scientific Chemicals

3-Formylthiophene-2-boronic acid, 97%, Thermo Scientific Chemicals

Catalog number: L15196.03
1 g, Each
Thermo Scientific Chemicals

3-Formylthiophene-2-boronic acid, 97%, Thermo Scientific Chemicals

Catalog number: L15196.03
1 g, Each
Quantity
Catalog number: L15196.03
also known as L15196-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17303-83-2
IUPAC Name
(3-formylthiophen-2-yl)boronic acid
Molecular Formula
C5H5BO3S
InChI Key
HYXMHAHVUFTVFZ-UHFFFAOYSA-N
SMILES
OB(O)C1=C(C=O)C=CS1
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥96.0%
Assay (HPLC)
≥96.0%
Proton NMR
Conforms to structure
Appearance (Color)
White to cream or pale yellow

Description

3-Formylthiophene-2-boronic acid is used as reactant for Suzuki-Miyaura cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Formylthiophene-2-boronic acid is used as reactant for Suzuki-Miyaura cross-coupling reactions.

Solubility
Slightly soluble in water.

Notes
Air Sensitive, store away form air. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text