Thermo Scientific Chemicals

Amberlite™ IRC120 Na ion exchange resin, Thermo Scientific Chemicals

Catalog number: L15199.0E
2500 g, Each
Thermo Scientific Chemicals

Amberlite™ IRC120 Na ion exchange resin, Thermo Scientific Chemicals

Catalog number: L15199.0E
2500 g, Each
Quantity
Catalog number: L15199.0E
also known as L15199-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78922-04-0
IUPAC Name
3-(3-chlorobenzenesulfonamido)benzoic acid
Molecular Formula
C13H10ClNO4S
InChI Key
APBOVLPLJFJSRI-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC(NS(=O)(=O)C2=CC(Cl)=CC=C2)=CC=C1
Form
Spherical beads
Appearance (Color)
Pale brown to brown
Comment
Max reversible swelling Na -> H ca. 10%
Exchange capacity
≥2.00 eq/L (Na+ form)
Matrix
Styrene divinylbenzene copolymer

Description

Amberlite™ IR-120(Na) ion exchange resin is used in water softening, dealkalinization and deionization. It is used for the recovery of toxic metals. It is also used in separation medium for TLC, paper chromatography and LPLC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amberlite™ IR-120(Na) ion exchange resin is used in water softening, dealkalinization and deionization. It is used for the recovery of toxic metals. It is also used in separation medium for TLC, paper chromatography and LPLC.

Solubility
Insoluble in water.

Notes
Water softening, dealkalinization, deionization
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text