Thermo Scientific Chemicals

3-Aminophthalhydrazide monosodium salt, 98+%, Thermo Scientific Chemicals

Catalog number: L15205.03
1 g, Each
Thermo Scientific Chemicals

3-Aminophthalhydrazide monosodium salt, 98+%, Thermo Scientific Chemicals

Catalog number: L15205.03
1 g, Each
Quantity
Catalog number: L15205.03
also known as L15205-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20666-12-0
IUPAC Name
sodium 8-amino-4-oxo-3,4-dihydrophthalazin-1-olate
Molecular Formula
C8H6N3NaO2
InChI Key
GBOXDRMCEVPUSV-UHFFFAOYSA-M
SMILES
[Na+].NC1=C2C([O-])=NNC(=O)C2=CC=C1
Assay (Aqueous acid-base Titration)
> 98.0%
Water Content (Karl Fischer Titration)
≤ 1.0%
Sulfated Ash
>34.95 to < 36.37% (theory 35.66%)
Form
powder
Appearance (Color)
White to cream or pale grey

Description

3-Aminophthalhydrazide monosodium salt is used for chemiluminescence analysis of metal cations, blood and glucococorticoids. It is also to image myeloperoxidase activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminophthalhydrazide monosodium salt is used for chemiluminescence analysis of metal cations, blood and glucococorticoids. It is also to image myeloperoxidase activity.

Solubility
Soluble in water (50 mg/ml).

Notes
Incompatible with bases, reducing agents and oxidizing agents. It is hygroscopic in nature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text