3-Aminophthalhydrazide monosodium salt is used for chemiluminescence analysis of metal cations, blood and glucococorticoids. It is also to image myeloperoxidase activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Aminophthalhydrazide monosodium salt is used for chemiluminescence analysis of metal cations, blood and glucococorticoids. It is also to image myeloperoxidase activity.
Solubility
Soluble in water (50 mg/ml).
Notes
Incompatible with bases, reducing agents and oxidizing agents. It is hygroscopic in nature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
General References:
- Yantu Zhang.; Zhujun Zhang.; Yonghua Song.; Yue Wei. Detection of glucocorticoid residues in pig liver by high-performance liquid chromatography with on-line electrogenerated [Cu(HIO6)2](5-)--luminol chemiluminescence detection. Journal of Chromatography A. 2007, 1154 (1-2), 260-268.
- Shimon Gross.; Seth T Gammon.; Britney L Moss.; Daniel Rauch.; John Harding.; Jay W Heinecke.; Lee Ratner.; David Piwnica-Worms. Bioluminescence imaging of myeloperoxidase activity in vivo. Nature Medicine. 2009, 15 (4), 455-461.