Thermo Scientific Chemicals

2-(4-Aminophenyl)ethanol, 97%, Thermo Scientific Chemicals

Catalog number: L15223.14
25 g, Each
Thermo Scientific Chemicals

2-(4-Aminophenyl)ethanol, 97%, Thermo Scientific Chemicals

Catalog number: L15223.14
25 g, Each
Quantity
Catalog number: L15223.14
also known as L15223-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
104-10-9
IUPAC Name
2-(4-aminophenyl)ethan-1-ol
Molecular Formula
C8H11NO
InChI Key
QXHDYMUPPXAMPQ-UHFFFAOYSA-N
SMILES
NC1=CC=C(CCO)C=C1
Appearance (Color)
Pale cream to cream to yellow to brown
Form
Crystals or powder or crystalline powder or lumps or granules
Assay (Non-aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (GC)
≥96.0%

Description

2-(4-Aminophenyl)ethanol is used as nonsymmetric monomer in the preparation of ordered [head-to-head (H-H) or tail-to-tail (T-T)] poly(amide-ester), in the synthesis of 4-aminostyrene and in functionalization of graphene nanoplatelets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(4-Aminophenyl)ethanol is used as nonsymmetric monomer in the preparation of ordered [head-to-head (H-H) or tail-to-tail (T-T)] poly(amide-ester), in the synthesis of 4-aminostyrene and in functionalization of graphene nanoplatelets.

Solubility
Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. It is sensitive to air. Incompatible with strong oxidizing agents. Store under dry inert gas. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text