2-(4-Aminophenyl)ethanol is used as nonsymmetric monomer in the preparation of ordered [head-to-head (H-H) or tail-to-tail (T-T)] poly(amide-ester), in the synthesis of 4-aminostyrene and in functionalization of graphene nanoplatelets.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(4-Aminophenyl)ethanol is used as nonsymmetric monomer in the preparation of ordered [head-to-head (H-H) or tail-to-tail (T-T)] poly(amide-ester), in the synthesis of 4-aminostyrene and in functionalization of graphene nanoplatelets.
Solubility
Soluble in methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. It is sensitive to air. Incompatible with strong oxidizing agents. Store under dry inert gas. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
- Li Li.; Koichiro Yonetake.; Osamu Haba.; Takeshi Endo.; Mitsuru Ueda. Constitutional Isomerism IV. Synthesis and Characterization of Poly(amide-ester)s from Isophthaloyl Chloride and 4-Aminophenethyl Alcohol. Polymer Journal. 2001, 33 (4), 364-70.
- Tokio Hagiwara.; Iwao Suzuki.; Kazushi Takeuchi.; Hiroshi Hamana.; Tadashi Narita. Synthesis and polymerization of N-(4-vinylphenyl)maleimide. Macromolecules. 1991, 24 (26), 6856-6858.