D-(-)-Quinic acid, 98%, Thermo Scientific Chemicals
D-(-)-Quinic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-(-)-Quinic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L15238.06
also known as L15238-06
Price (USD)/ Each
33.50
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
33.50
Add to cart
D-(-)-Quinic acid, 98%, Thermo Scientific Chemicals
Catalog numberL15238.06
Price (USD)/ Each
33.50
-
Add to cart
Chemical Identifiers
CAS77-95-2
IUPAC Name1,3,4,5-tetrahydroxycyclohexane-1-carboxylic acid
Molecular FormulaC7H12O6
InChI KeyAAWZDTNXLSGCEK-UHFFFAOYNA-N
SMILESOC1CC(O)(CC(O)C1O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystalline powder
Identification (FTIR)Conforms
Optical Rotation-42° to -45° (c=10 in water)
Melting Point (clear melt)158-168°C
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
View more
D(-)-Quinic Acid is used in the synthesis of anti-influenza/anti-swine flu medication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D(-)-Quinic Acid is used in the synthesis of anti-influenza/anti-swine flu medication.

Solubility
Soluble in water (400g/L), methanol and dimethyl sulfoxide.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Timothy J Key et. al. Plasma carotenoids, retinol, and tocopherols and the risk of prostate cancer in the European Prospective Investigation into Cancer and Nutrition study. American Journal of Clinical Nutrition.2007, 86,(3), 672-681.
  2. Nadine Volz et. al. Effect of coffee combining green coffee bean constituents with typical roasting products on the Nrf2/ARE pathway in vitro and in vivo. Journal of Agricultural and Food Chemistry.2012, 60, (38), 9631-9641 .
  3. Chiral starting material, used for example in the synthesis of the angucycline antibiotics urdamycinone B and 104-2: J. Am. Chem. Soc., 117, 8472 (1995).