Thermo Scientific Chemicals

D-(-)-Quinic acid, 98%, Thermo Scientific Chemicals

Catalog number: L15238.14
25 g, Each
Thermo Scientific Chemicals

D-(-)-Quinic acid, 98%, Thermo Scientific Chemicals

Catalog number: L15238.14
25 g, Each
Quantity
Catalog number: L15238.14
also known as L15238-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77-95-2
IUPAC Name
1,3,4,5-tetrahydroxycyclohexane-1-carboxylic acid
Molecular Formula
C7H12O6
InChI Key
AAWZDTNXLSGCEK-UHFFFAOYNA-N
SMILES
OC1CC(O)(CC(O)C1O)C(O)=O
Form
Powder or crystalline powder
Identification (FTIR)
Conforms
Optical Rotation
-42° to -45° (c=10 in water)
Melting Point (clear melt)
158-168°C
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

D(-)-Quinic Acid is used in the synthesis of anti-influenza/anti-swine flu medication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D(-)-Quinic Acid is used in the synthesis of anti-influenza/anti-swine flu medication.

Solubility
Soluble in water (400g/L), methanol and dimethyl sulfoxide.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text