(+/-)-1-Amino-2-propanol, 94%, remainder 2-amino-1-propanol, Thermo Scientific Chemicals
(+/-)-1-Amino-2-propanol, 94%, remainder 2-amino-1-propanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(+/-)-1-Amino-2-propanol, 94%, remainder 2-amino-1-propanol, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
2500 g
Catalog number L15343.30
also known as L15343-30
Price (USD)/ Each
22.50
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
22.50
Add to cart
(+/-)-1-Amino-2-propanol, 94%, remainder 2-amino-1-propanol, Thermo Scientific Chemicals
Catalog numberL15343.30
Price (USD)/ Each
22.50
-
Add to cart
Chemical Identifiers
CAS78-96-6
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥92.0%
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Refractive Index1.4460-1.4500 @ 20?C
Amino-2-propanol is used to study the effect of temperature and water content on the molecular structure and hydrogen bonding of amino-2-propanol, which has been examined by Fourier transform near-infrared (FT-NIR) spectroscopy. Also used in used in the synthesis of protein kinase CK2 inhibitors used in the treatment of neoplasia and other infective diseases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Fully miscible with water.

Notes
Air Sensitive and Hygroscopic, store away form air and water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Krzysztof Zdzisław Haufa; and Mirosław Antoni Czarnecki. Molecular structure and hydrogen bonding of 2-aminoethanol, 1-amino-2-propanol, 3-amino-1-propanol, and binary mixtures with water studied by Fourier transform near-infrared spectroscopy and density functional theory calculations. Applied Spectroscopy.2010, 64,(3), 351-358.
  2. Tomáš Etrych et. al. Polymer conjugates of doxorubicin bound through an amide and hydrazone bond: Impact of the carrier structure onto synergistic action in the treatment of solid tumours. European Journal of Pharmaceutical Sciences.2012, 58, (38), 1-12.