N-Benzylglycine, 98+%, Thermo Scientific Chemicals
N-Benzylglycine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Benzylglycine, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L15368.14
also known as L15368-14
Price (USD)/ Each
218.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
218.00
Add to cart
N-Benzylglycine, 98+%, Thermo Scientific Chemicals
Catalog numberL15368.14
Price (USD)/ Each
218.00
-
Add to cart
Chemical Identifiers
CAS17136-36-6
IUPAC Name2-(benzylamino)acetic acid
Molecular FormulaC9H11NO2
InChI KeyKGSVNOLLROCJQM-UHFFFAOYSA-N
SMILESOC(=O)CNCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
FormCrystals or powder or crystalline powder or fused solid
Appearance (Color)White to cream to pale brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only