Thermo Scientific Chemicals

L-Pipecolinic acid, 99%, Thermo Scientific Chemicals

Catalog number: L15373.06
5 g, Each
Thermo Scientific Chemicals

L-Pipecolinic acid, 99%, Thermo Scientific Chemicals

Catalog number: L15373.06
5 g, Each
Quantity
Catalog number: L15373.06
also known as L15373-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3105-95-1
IUPAC Name
piperidine-2-carboxylic acid
Molecular Formula
C6H11NO2
InChI Key
HXEACLLIILLPRG-UHFFFAOYNA-N
SMILES
OC(=O)C1CCCCN1
Form
Powder
Optical Rotation
-26.6 ± 0.5? (c=1 in water)
Appearance (Color)
White to cream
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

L-Pipecolinic acid is involved in synaptic transmission in the central nervous system. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Pipecolinic acid is involved in synaptic transmission in the central nervous system. It is also used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text