Diisobutylamine was used to study the effect of achiral amine on hydrogenation of ethyl pyruvate over cinchonidine-Pt/Al2O3 catalyst system.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diisobutylamine was used to study the effect of achiral amine on hydrogenation of ethyl pyruvate over cinchonidine-Pt/Al2O3 catalyst system.
Solubility
Soluble in water 5 g/L (20°C).
Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
RUO – Research Use Only
General References:
- Tálas E, et al. Influence of achiral amine additives on the Orito's reaction. J. Mol. Catal. A: Chem. 2012, 357 87-94.
- Ivanov AV, et al. Adducts of zinc and copper (II) dialkyldithiocarbamate complexes with dibutyl-and diisobutylamines: Synthesis, structures, EPR and solid-state natural abundance 13C and 15N CP/MAS NMR spectra. Russ. J. Coord. Chem. 2007, 33 (4), 233-243.