Thermo Scientific Chemicals

Diisobutylamine, 99%, Thermo Scientific Chemicals

Catalog number: L15495.AU
1000 mL, Each
Thermo Scientific Chemicals

Diisobutylamine, 99%, Thermo Scientific Chemicals

Catalog number: L15495.AU
1000 mL, Each
Quantity
Catalog number: L15495.AU
also known as L15495-AU
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-96-3
Refractive Index
1.4070-1.4110 @ 20?C
Form
Liquid
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%

Description

Diisobutylamine was used to study the effect of achiral amine on hydrogenation of ethyl pyruvate over cinchonidine-Pt/Al2O3 catalyst system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diisobutylamine was used to study the effect of achiral amine on hydrogenation of ethyl pyruvate over cinchonidine-Pt/Al2O3 catalyst system.

Solubility
Soluble in water 5 g/L (20°C).

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text