2,3-Dibromofuran, 97%, stab. with 0.5% calcium carbonate, Thermo Scientific Chemicals
2,3-Dibromofuran, 97%, stab. with 0.5% calcium carbonate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3-Dibromofuran, 97%, stab. with 0.5% calcium carbonate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L15518.06
also known as L15518-06
Price (USD)/ Each
175.65
Online exclusive
195.00 
Save 19.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
175.65
Online exclusive
195.00 
Save 19.35 (10%)
Add to cart
2,3-Dibromofuran, 97%, stab. with 0.5% calcium carbonate, Thermo Scientific Chemicals
Catalog numberL15518.06
Price (USD)/ Each
175.65
Online exclusive
195.00 
Save 19.35 (10%)
-
Add to cart
Chemical Identifiers
CAS30544-34-4
IUPAC Name2,3-dibromofuran
Molecular FormulaC4H2Br2O
InChI KeyGKPGEBCMRMQOPF-UHFFFAOYSA-N
SMILESBrC1=C(Br)C=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Appearance (Color)Clear to turbid colorless to yellow or pale brown
Refractive Index1.5460-1.5510 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only