Thermo Scientific Chemicals

1-Ethylpiperazine, 98%, Thermo Scientific Chemicals

Catalog number: L15539.36
500 g, Each
Thermo Scientific Chemicals

1-Ethylpiperazine, 98%, Thermo Scientific Chemicals

Catalog number: L15539.36
500 g, Each
Quantity
Catalog number: L15539.36
also known as L15539-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5308-25-8
IUPAC Name
1-ethylpiperazine
Molecular Formula
C6H14N2
InChI Key
WGCYRFWNGRMRJA-UHFFFAOYSA-N
SMILES
CCN1CCNCC1
Identification (FTIR)
Conforms
Assay (GC)
≥97.5%
Refractive Index
1.4670-1.4710 @ 20?C
Appearance (Color)
Clear colorless to yellow
Form
Liquid

Description

1-Ethylpiperazine is used in the synthesis of 2-(2-methoxy-5-((4-methylpiperazin-1-yl)sulfonyl)phenyl)-1H-benzo[d]imidazole hydrochloride and 2-(5-((4-ethylpiperazin-1-yl)sulfonyl)-2-methoxyphenyl)-1H-benzo[d]imidazole hydrochloride. It is also employed as an intermediate in veterinary medicine, especially in the production of ciproflmoxacin ethyl. Further, it serves as a precursor to prepare dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethylpiperazine is used in the synthesis of 2-(2-methoxy-5-((4-methylpiperazin-1-yl)sulfonyl)phenyl)-1H-benzo[d]imidazole hydrochloride and 2-(5-((4-ethylpiperazin-1-yl)sulfonyl)-2-methoxyphenyl)-1H-benzo[d]imidazole hydrochloride. It is also employed as an intermediate in veterinary medicine, especially in the production of ciproflmoxacin ethyl. Further, it serves as a precursor to prepare dyes.

Solubility
Miscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text