Thermo Scientific Chemicals

4-Hydroxybenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: L15594.06
5 g, Each
Thermo Scientific Chemicals

4-Hydroxybenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: L15594.06
5 g, Each
Quantity
Catalog number: L15594.06
also known as L15594-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71597-85-8
IUPAC Name
(4-hydroxyphenyl)boronic acid
Molecular Formula
C6H7BO3
InChI Key
COIQUVGFTILYGA-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=C(O)C=C1
Assay (Aqueous acid-base Titration)
≥96.0%
Proton NMR
Conforms to structure
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder or granules
Assay (HPLC)
≥96.0%

Description

4-Hydroxybenzeneboronic acid is used for Suzuki-Miyaura coupling and Stille coupling and potential for introducing different alkyl groups, palladium-catalyzed aminocarbonylation and cross-coupling reactions, bio-supported palladium nanoparticles as phosphine-free catalyst for Suzuki reaction in water and Cu2O-catalyzed aerobic oxidative cross-coupling of tetrazoles. It is also used in preparation of rod-like dendronized polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybenzeneboronic acid is used for Suzuki-Miyaura coupling and Stille coupling and potential for introducing different alkyl groups, palladium-catalyzed aminocarbonylation and cross-coupling reactions, bio-supported palladium nanoparticles as phosphine-free catalyst for Suzuki reaction in water and Cu2O-catalyzed aerobic oxidative cross-coupling of tetrazoles. It is also used in preparation of rod-like dendronized polymers.

Solubility
Soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text