Thermo Scientific Chemicals

Eaton's Reagent, P{2}O{5} ca 7.5% w/w

Catalog number: L15607.AP
500 mL, Each
Thermo Scientific Chemicals

Eaton's Reagent, P{2}O{5} ca 7.5% w/w

Catalog number: L15607.AP
500 mL, Each
Quantity
Catalog number: L15607.AP
also known as L15607-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39394-84-8
IUPAC Name
diphosphooxidane; methanesulfonic acid
Molecular Formula
CH4O8P2S
InChI Key
JHNLZOVBAQWGQU-UHFFFAOYSA-N
SMILES
CS(O)(=O)=O.O=P(=O)OP(=O)=O
Refractive Index
1.4325-1.4375 @ 20?C
Assay (Aqueous acid-base Titration)
≥6.5 to ≤10.0% w/w
Appearance (Color)
Clear colorless to yellow to orange to pale brown or pale pink
Comment
May contain precipitate: Re-dissolves on warming.
Form
Solution

Description

Eaton's Reagent is used in the synthesis fluorine-based hole-transporting material, mono and bis-chalcone derivatives via Claisen-Schmidt condensation reaction between arylaldehydes and ketones and 4-hydroxycoumarins and 4-hydroxy-2-quinolinones derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text