2,3-Diphenylmaleic anhydride, 98%, Thermo Scientific Chemicals
2,3-Diphenylmaleic anhydride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3-Diphenylmaleic anhydride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L15612.03
also known as L15612-03
Price (USD)/ Each
52.60
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
52.60
Add to cart
2,3-Diphenylmaleic anhydride, 98%, Thermo Scientific Chemicals
Catalog numberL15612.03
Price (USD)/ Each
52.60
-
Add to cart
Chemical Identifiers
CAS4808-48-4
IUPAC Namediphenyl-2,5-dihydrofuran-2,5-dione
Molecular FormulaC16H10O3
InChI KeyOUJCFCNZIUTYBH-UHFFFAOYSA-N
SMILESO=C1OC(=O)C(=C1C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)154.5-160.5?C
Appearance (Color)Yellow to brown
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only