Thermo Scientific Chemicals

2,2-Diphenylpropylamine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: L15615.03
1 g, Each
Thermo Scientific Chemicals

2,2-Diphenylpropylamine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: L15615.03
1 g, Each
Quantity
Catalog number: L15615.03
also known as L15615-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
40691-66-5
IUPAC Name
hydrogen 2,2-diphenylpropan-1-amine chloride
Molecular Formula
C15H18ClN
InChI Key
AASCJSPDUDWGGQ-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CC(CN)(C1=CC=CC=C1)C1=CC=CC=C1
Form
Powder
Assay (Titration ex Chloride)
≥98.0 to ≤102.0%
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text