Thermo Scientific Chemicals

4,4'-Oxybis(benzoic acid), 98+%, Thermo Scientific Chemicals

Catalog number: L15651.22
100 g, Each
Thermo Scientific Chemicals

4,4'-Oxybis(benzoic acid), 98+%, Thermo Scientific Chemicals

Catalog number: L15651.22
100 g, Each
Quantity
Catalog number: L15651.22
also known as L15651-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2215-89-6
IUPAC Name
4-(4-carboxyphenoxy)benzoic acid
Molecular Formula
C14H10O5
InChI Key
WVDRSXGPQWNUBN-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(OC2=CC=C(C=C2)C(O)=O)C=C1
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Assay (HPLC)
≥98.0%
Water Content (Karl Fischer Titration)
≤1.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text