Thermo Scientific Chemicals

2,3-Dimethylfuran, 99%, Thermo Scientific Chemicals

Catalog number: L15705.06
5 g, Each
Thermo Scientific Chemicals

2,3-Dimethylfuran, 99%, Thermo Scientific Chemicals

Catalog number: L15705.06
5 g, Each
Quantity
Catalog number: L15705.06
also known as L15705-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14920-89-9
IUPAC Name
2,3-dimethylfuran
Molecular Formula
C6H8O
InChI Key
FJSKXQVRKZTKSI-UHFFFAOYSA-N
SMILES
CC1=C(C)C=CO1
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.4420-1.4450 @ 20?C
Appearance (Color)
Clear colorless to yellow
Form
Liquid

Description

2,3-Dimethylfuran may be used in the synthesis of 2,9-dioxabicyclo[3.3.1]nonane. Ireland Alcohol was synthesized in from 2,3-dimethylfuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dimethylfuran may be used in the synthesis of 2,9-dioxabicyclo[3.3.1]nonane. Ireland Alcohol was synthesized in from 2,3-dimethylfuran.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text