Thermo Scientific Chemicals

N-Methyldiethanolamine, 98+%, Thermo Scientific Chemicals

Catalog number: L15712.22
100 g, Each
Thermo Scientific Chemicals

N-Methyldiethanolamine, 98+%, Thermo Scientific Chemicals

Catalog number: L15712.22
100 g, Each
Quantity
Catalog number: L15712.22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
105-59-9
IUPAC Name
2-[(2-hydroxyethyl)(methyl)amino]ethan-1-ol
Molecular Formula
C5H13NO2
InChI Key
CRVGTESFCCXCTH-UHFFFAOYSA-N
SMILES
CN(CCO)CCO
Comment
Purchased in U.K. and U.S.
Refractive Index
1.4675-1.4715 @20°C (U.K. specification)
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0% (U.K. specification)

Description

N-Methyldiethanolamine is a reagent used for protection of boronic acids as N-methyl-O,O-diethanolamine esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methyldiethanolamine is a reagent used for protection of boronic acids as N-methyl-O,O-diethanolamine esters.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text