Thermo Scientific Chemicals

Hexafluorophosphoric acid, ca 60% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: L15728.AC
25 mL, Each
Thermo Scientific Chemicals

Hexafluorophosphoric acid, ca 60% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: L15728.AC
25 mL, Each
Quantity
Catalog number: L15728.AC
also known as L15728-AC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16940-81-1
IUPAC Name
hydrogen hexafluoro-λ⁵-phosphanuide
Molecular Formula
F6HP
InChI Key
LJQLCJWAZJINEB-UHFFFAOYSA-O
SMILES
[H+].F[P-](F)(F)(F)(F)F
Assay (unspecified)
55-65%
Appearance (Color)
Clear, colorless to pale pink or pale yellow
Form
Liquid

Description

Hexafluorophosphoric acid is used as a catalyst. It is also used as a metal cleaner and chemical polishing agent for the formation of protective coatings of metal surfaces..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexafluorophosphoric acid is used as a catalyst. It is also used as a metal cleaner and chemical polishing agent for the formation of protective coatings of metal surfaces..

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with strong bases, glass, alkali metals, powdered metals, strong oxidizing agents, metals and reacts violently with water.  
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text