Thermo Scientific Chemicals

Tri-n-octylphosphine, tech. 90%, Thermo Scientific Chemicals

Catalog number: L15744.AP
500 mL, Each
Thermo Scientific Chemicals

Tri-n-octylphosphine, tech. 90%, Thermo Scientific Chemicals

Catalog number: L15744.AP
500 mL, Each
Quantity
Catalog number: L15744.AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4731-53-7
IUPAC Name
trioctylphosphane
Molecular Formula
C24H51P
InChI Key
RMZAYIKUYWXQPB-UHFFFAOYSA-N
SMILES
CCCCCCCCP(CCCCCCCC)CCCCCCCC
Form
Liquid
Assay from Supplier's CofA
≥88.0%
Comment
May become cloudy on storage and form precipitate on standing

Description

Tri-n-octylphosphine is used for coating zinc sulfide shells on cadmium-selenium quantum dot core by successive ionic layer adsorption and reaction method. It acts as a precursor to trioctylphosphine oxide. Further, it is used as a solvent for cadmium and selenium precursors. It also serves as a common reagent in the chemical synthesis of nanoparticles. It acts as a precursor to trioctylphosphine oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-octylphosphine is used for coating zinc sulfide shells on cadmium-selenium quantum dot core by successive ionic layer adsorption and reaction method. It acts as a precursor to trioctylphosphine oxide. Further, it is used as a solvent for cadmium and selenium precursors. It also serves as a common reagent in the chemical synthesis of nanoparticles. It acts as a precursor to trioctylphosphine oxide.

Solubility
Immiscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text